Skip to Content
Merck
All Photos(1)

Key Documents

QC1452

Supelco

Total Petroleum Hydrocarbons (TPH)

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC QCI-1377

concentration

in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
food and beverages

format

matrix material

storage temp.

2-30°C

General description

This product has been discontinued but product QC1800-2ML may be an acceptable replacement.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup - Solvent

Flash Point(C)

-17 °C - closed cup - Solvent


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of sample matrix on the determination of total petroleum hydrocarbons (TPH) in soil by gas chromatography?flame ionization detection.
Saari E, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 87(2), 113-118 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service