Skip to Content
Merck
All Photos(1)

Key Documents

QC1194

Supelco

Settleable Solids - WP

certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€55.40

€55.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
€55.40

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€55.40


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

packaging

jar of 30 mL

manufacturer/tradename

RTC QCI-253

concentration

(NaCl)

matrix

NaCl

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service