PSRVD02
Multi-Parameter Refractive Index & Density Certified Reference Material at 15 °C, 20 °C and 25 °C, Single 30 mL
Nominal RI Value 1.4217 at 20 °C
About This Item
Recommended Products
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- CRM certified in strict adherence to ISO 17025 / ISO 17034 under UKAS accreditation
- density measured and qualified following ASTM D1480
- ensures full compliance to ASTM and IP test method protocols
- low levels of uncertainty, guaranteeing maximum accuracy of data and dependable results
- supplied in 30 mL volume sealed glass vials for ample measurement readings
- 12-months shelf life as stated in the certificate of analysis (CoA)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1
Supplementary Hazards
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
158.0 °F - Pensky-Martens closed cup
Flash Point(C)
70 °C - Pensky-Martens closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service