Skip to Content
Merck
All Photos(4)

Key Documents

PPB008

Sigma-Aldrich

Tris Acetate-EDTA buffer

pH 8.3, pHast Pack, powder

Synonym(s):

1X TAE buffer, TAE, Tris Acetate EDTA, TAE buffer

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
€94.50

€94.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 PKG
€94.50

About This Item

MDL number:
UNSPSC Code:
41105319
NACRES:
NA.21

€94.50


Please contact Customer Service for Availability

Request a Bulk Order

product line

pHast Pack

form

powder

pH

8.3

suitability

suitable for gel electrophoresis (after dilution to working concentration)
suitable for gel electrophoresis

foreign activity

DNAse, none detected
Nickase, none detected
Protease, none detected
RNAse, none detected

SMILES string

CC(O)=O.NC(CO)(CO)CO.OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8.C4H11NO3.C2H4O2/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;5-4(1-6,2-7)3-8;1-2(3)4/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);6-8H,1-3,5H2;1H3,(H,3,4)

InChI key

HGEVZDLYZYVYHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris Acetate-EDTA (TAE) buffer, with a pH range of 8.1 – 8.4, is typically used for agarose gel electrophoresis, especially for the recovery of DNA. For larger double-stranded DNA fragments (>12kB) TAE buffer is preferred. However, the buffering capacity can become exhausted if electrophoresis time is extended. Buffer circulation or replacement during electrophoresis can remedy the lower buffering capacity.

Application

Suitable as a running and gel preparation buffer in:
  • DNA agarose gel electrophoresis
  • Non-denaturing RNA agarose gel electrophoresis for RNA >1500 bases
  • Denaturing gradient gel electrophoresis
  • Northern and Southern blotting
TAE running buffer is the most commonly used buffer for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis. Double-stranded DNA tends to run faster in TAE than in other buffers but can also become exhausted during extended electrophoresis. Buffer circulation or buffer replacement during extended electrophoresis can remedy the lower buffering capacity. Dilution of the concentrated TAE buffer produces a 1× TAE buffer with 40 mM Tris-acetate and 1 mM EDTA, pH 8.3. The 1× TAE buffer is used both in the agarose gel and as a running buffer. Applied voltages of less than 5 V/cm (distance between the electrodes of the unit) are recommended for maximum resolution.

Features and Benefits

  • Ready for use – saves time and effort
  • No measuring and pH adjusting needed
  • Eliminate exposure to toxic chemicals to prepare buffers
  • Biological tests: free of DNase, RNase, Protease, and Nickase
  • Chemical tests: Iron ≤10 ppm, lead ≤5 ppm

Packaging

Foil pouches

Preparation Note

Prepared with Biotechnology Performance Certified Trizma base (T6066) and Molecular Biology Reagent EDTA disodium salt (E5134). Solutions also contain acetic acid (A6283); powdered blend contains Trizma acetate (T1258).

Reconstitution

Contents of one pouch, when dissolved in 500mL of distilled or deionized water, will yield a 1X solution containing 40mM Tris-Acetate, 1mM EDTA, pH 8.3 at 25 °C. Certified to be DNAse, RNAse, Protease, and Nickase free. Certified for use in electrophoresis.

Storage and Stability

Store at room temperature. Product may naturally agglomerate but can be simply broken up within the pouch prior to use.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

pHast Pack is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service