Skip to Content
Merck
All Photos(1)

Key Documents

PHR1765

Supelco

Cycloserine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

D-Cycloserine, (R)-4-Amino-3-isoxazolidone, 4-Amino-3-isoxazolidinone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€137.00

€137.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€137.00

About This Item

Empirical Formula (Hill Notation):
C3H6N2O2
CAS Number:
Molecular Weight:
102.09
Beilstein:
80798
EC Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.24

€137.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1158005

API family

cycloserine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

mp

147 °C (dec.) (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

-10 to -25°C

SMILES string

N[C@@H]1CONC1=O

InChI

1S/C3H6N2O2/c4-2-1-7-5-3(2)6/h2H,1,4H2,(H,5,6)/t2-/m1/s1

InChI key

DYDCUQKUCUHJBH-UWTATZPHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cycloserine is a broad-spectrum antibiotic commonly produced by certain strains of Streptomyces orchidaceus or S. garphalus.[1] It is used as a second-line drug treatment for multiple drug-resistant strains of Mycobacterium tuberculosis.[2][3]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Cycloserine may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by high performance liquid chromatography.[3][4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Mode of Action: Inhibits cell wall biosynthesis (D-Ala peptide bond formation). Also prevents conversion of D-Ala to L-Ala. Bacteriostatic.
Partial agonist at the glycine modulatory site of NMDA glutamatergic receptors; antibiotic against Gram-negative bacteria.
Mode of Resistance: D-Ala transport interference.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8432 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development and validation of indirect RP-HPLC method for enantiomeric purity determination of d-cycloserine drug substance
Karthikeyan K, et al.
Journal of Pharmaceutical and Biomedical Analysis, 54(4), 850-854 (2011)
d-Cycloserine
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 457-462 (2015)
Development of a liquid chromatographic method for the determination of related substances and assay of d-cycloserine
Pendela M, et al.
Journal of Pharmaceutical and Biomedical Analysis, 47(4-5), 807-811 (2008)
S David
The Journal of antimicrobial chemotherapy, 47(2), 203-206 (2001-02-07)
D-Cycloserine (DCS) is a peptidoglycan inhibitor. Although very effective against Mycobacterium tuberculosis, it is seldom employed in the management of this infection due to its high toxicity. beta-Chloro-D-alanine (another peptidoglycan inhibitor) reduces the MIC of DCS from 50 to 2.5

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service