Skip to Content
Merck
All Photos(1)

Key Documents

PHR1029

Supelco

Tocopheryl Acid Succinate, a

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

D-α-Tocopherol succinate, Vitamin E succinate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€88.40

€88.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€88.40

About This Item

Empirical Formula (Hill Notation):
C33H54O5
CAS Number:
Molecular Weight:
530.78
Beilstein:
4038233
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€88.40


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. T1610000
traceable to USP 1667803

API family

tocopherol

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(=O)CCC(O)=O)c(C)c(C)c2O1

InChI

1S/C33H54O5/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-20-33(8)21-19-28-27(7)31(25(5)26(6)32(28)38-33)37-30(36)18-17-29(34)35/h22-24H,9-21H2,1-8H3,(H,34,35)/t23-,24-,33-/m1/s1

InChI key

IELOKBJPULMYRW-NJQVLOCASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards. Tocopheryl Acid Succinate, a is a derivative of vitamin E and a potent and highly specific anticancer agent known to inhibit the growth of cancer cells.[1]

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3258 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin E succinate induces Fas-mediated apoptosis in estrogen receptor-negative human breast cancer cells
Turley JM, et al.
Cancer Research, 57(5), 881-890 (1997)
Pengfei Xu et al.
Biomaterials, 35(26), 7574-7587 (2014-06-06)
Metastasis is one of the major obstacles for the successful therapy of breast cancer. Although increased candidate drugs targeting cancer metastasis are tested, their clinical translation is limited by either serve toxicity or low efficacy. In present work, a nano-drug
Sindhuri Maddineni et al.
AAPS PharmSciTech, 16(2), 444-454 (2014-11-02)
The objective of the present study was to investigate the effects of processing variables and formulation factors on the characteristics of hot-melt extrudates containing a copolymer (Kollidon® VA 64). Nifedipine was used as a model drug in all of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service