Skip to Content
Merck
All Photos(1)

Key Documents

P57506-M

Sigma-Aldrich

Pyridine

reagent grade, ≥99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€226.00
2.5 L
€770.00

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€226.00
2.5 L
€770.00

About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
Beilstein:
103233
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
application(s):
peptide synthesis
bp:
115 °C (lit.)
vapor pressure:
10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

reagent grade

vapor density

2.72 (vs air)

vapor pressure

10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

Assay

≥99%

form

liquid

autoignition temp.

899 °F

expl. lim.

12.4 %

impurities

≤0.1% water (Karl Fischer)

refractive index

n20/D 1.509 (lit.)

pH

8.5 (25 °C, 15.82 g/L)

bp

115 °C (lit.)

mp

−42 °C (lit.)

density

0.978 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

C1=CN=CC=C1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridine is a basic N-heterocyclic compound. It acts as a nitrogen donor ligand and forms many metal-pyridine complexes. It has wide applications as a solvent, catalyst, and intermediate for synthesis.[1]

Application


  • Ligand-Modulated Nuclearity and Geometry in Nickel(II) Hydrazone Complexes: From Mononuclear Complexes to Acetato- and/or Phenoxido-Bridged Clusters.: Research on the use of pyridine and its derivatives to control the nuclearity and coordination geometry in nickel(II) complexes, which is crucial for designing metal-organic frameworks and catalytic sites (Vrdoljak et al., 2023).

  • Non-palindromic (C^C^D) gold(III) pincer complexes are not accessible by intramolecular oxidative addition of biphenylenes - an experimental and quantum chemical study.: A study examining the limitations of pyridine-containing ligands in stabilizing reactive gold(III) complexes, providing insights into the electronic properties and stability challenges (Feuerstein & Breher, 2021).


Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biotreatment of waste gas containing pyridine in a biofilter
Pandey RA, et al.},
Bioresource Technology, 98(12), 2258-2267 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service