Skip to Content
Merck
All Photos(1)

Key Documents

P2182

Millipore

Potato Dextrose Agar

Recommended for the isolation of yeasts and molds, NutriSelect® Basic

Synonym(s):

PDA

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€110.00
1 KG
€372.00

€110.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
€110.00
1 KG
€372.00

About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

€110.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

potato

Quality Level

sterility

non-sterile

form

powder

manufacturer/tradename

NutriSelect® Basic

pH

-5.6-25 °C

application(s)

bioburden testing
cosmetics
food and beverages
microbiology
pharmaceutical
sterility testing

storage temp.

10-30°C

suitability

bacteria
molds
yeasts

General description

Potato Dextrose Agar is used for the cultivation of bacteria, fungi, yeast, and molds. The low pH makes the media selective for acid tolerant bacteria, fungi, mold, and yeasts. It is used for studying ascospore dispersal. Potato extract serves as a source of carbon, nitrogen, minerals, vitamins, and other essential growth nutrients. Dextrose acts as source of carbohydrate.

Application

PDA is recommended for use in: -
  • The isolation and enumeration of yeasts and molds from food and dairy products
  • Stimulating sporulation
  • Maintaining stock cultures of fungi
  • Differentiating various dermatophytes based on pigment production

Components

Potato extract - 4 g/L
Dextrose - 20 g/L
Agar - 15 g/L

Other Notes

Contains: Infusions from potatoes, glucose and agar.

Preparation Note

Solve 39 g in 1 litre distilled water. Autoclave 15 minutes at 121°C.

Analysis Note

Microbiologically tested.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sclerotinia Diseases of Crop Plants: Biology, Ecology and Disease Management, 403-403 (2008)
Two convenient methods to evaluate soybean for resistance to Sclerotinia sclerotiorum
Chen Y and Wang D
Plant Disease, 89(12), 1268-1272 (2005)
Effect of chestnut tannin extract (Castanea sativa Miller) on the proliferation of Cladosporium cladosporioides on sheep cheese rind during the ripening
Messini A, et al.
International dairy journal, 66, 6-12 (2017)
Julia Schumacher et al.
Eukaryotic cell, 7(4), 584-601 (2008-02-12)
Recently, we showed that the alpha subunit BCG1 of a heterotrimeric G protein is an upstream activator of the Ca(2+)/calmodulin-dependent phosphatase calcineurin in the gray mold fungus Botrytis cinerea. To identify the transcription factor acting downstream of BCG1 and calcineurin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service