Skip to Content
Merck
All Photos(1)

Key Documents

NISTRM8631B

Sigma-Aldrich

Medium Test Dust (MTD)

NIST® RM 8631b, pkg of 20 g

Sign Into View Organizational & Contract Pricing

Select a Size

20 G
€784.00

€784.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 G
€784.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€784.00


Please contact Customer Service for Availability

Request a Bulk Order

packaging

pkg of 20 g

manufacturer/tradename

NIST® RM 8631b

application(s)

environmental

General description

A unit of Reference Material (RM) 8631b, an ISO Medium Test Dust (MTD), consists of 20 g of a natural mineral dust that is heterogeneous in composition and polydisperse with respect to size. RM 8631b is intended to be used as a secondary material for calibrating particle sizing instruments, especially optical particle counters, when used in conjunction with published standard methods.

SRM 8631B_cert

SRM 8631B_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service