Skip to Content
Merck
All Photos(1)

Key Documents

NISTRM8496

Eucalyptus hardwood

NIST® RM 8496, bleached kraft pulp

Synonym(s):

Cellulose pulp

Sign Into View Organizational & Contract Pricing

Select a Size

10 SHEETS
€404.00

€404.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 SHEETS
€404.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€404.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service