Skip to Content
Merck
All Photos(1)

Key Documents

NIST8851

Zeolite A

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

35 G
€642.00

€642.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
35 G
€642.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€642.00


Please contact Customer Service for Availability

Request a Bulk Order

form

solid

packaging

pkg of 35-40 g

manufacturer/tradename

NIST®

SMILES string

[Si](=O)=O.[Al](=O)O[Al]=O

InChI

1S/2Al.O2Si.3O/c;;1-3-2;;;

InChI key

HNPSIPDUKPIQMN-UHFFFAOYSA-N

General description

Each unit of RM contains ~ 35 g to 40 g of material enclosed in a sealed container.

SRM 8851_cert

SRM 8851_SDS

Application

The Reference Material (RM) is intended for use as a Zeolite A source for measurement comparisons.

Features and Benefits

  • Available with the certificate documenting NIST-certified reference mass fraction values for components of Zeolite A.
  • Information values for atomic ratio of Si to Al is provided.
  • The NIST certificate is provided with expiration certificate, storage, handling, use, and maintenance instructions.

Other Notes

Reference/Informational value is provided for the following analytes.
Sodium (Na), Aluminum(Al), Silicon (Si), Iron (Fe), Cerium (Ce), Cobalt (Co), Chromium (Cr), Cesium (Cs), Europium (Eu), Hafnium (Hf), Lanthanum (La), Scandium (Sc), Samarium (Sm), Thorium (Th), Zinc (Zn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service