Skip to Content
Merck
All Photos(1)

Key Documents

NIST3278

Tocopherols in edible oils

NIST® SRM® 3278

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 1 ML
€842.00

€842.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 X 1 ML
€842.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€842.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1 mL

manufacturer/tradename

NIST®

format

matrix material

General description

One unit of SRM 3278 contains five oil ampoules, each containing about 1 mL of material under an argon atmosphere.

SRM 3278_cert

SRM 3278_SDS

Application

Tocopherols in edible oils is intended for use inthe validation of analytical methods to determine tocopherols in edible oils and similar matrices. Additionally, it is suitable for quality assurance purposes when assigning values to in-house control materials.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction values of tocopherols in SRM 3278.
  • The certified value is metrologically traceable to the SI unit of mass fraction, expressed as micrograms per gram.
  • The NIST certificate is provided with expiration certificate, storage, handling, use,and maintenance instructions.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Vitamins: α-Tocopherol, β-Tocopherol, γ-Tocopherol, δ-Tocopherol

Matrix Group: Nuts, Soy, Edible Oils and Fats

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service