Skip to Content
Merck
All Photos(1)

Key Documents

NIST3180

Iodide anion (I-) standard solution

NIST® SRM® 3180

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 5 ML
€1,590.00

€1,590.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 X 5 ML
€1,590.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€1,590.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

solution

packaging

pkg of 5 x 5 mL

manufacturer/tradename

NIST®

General description

This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of iodide. A unit of SRM 3180 consists of five 5 mL, sealed, amber, borosilicate glass ampoules of solution prepared gravimetrically to contain a known mass fraction of iodide. The matrix of the solution is water containing sodium sulfite at a concentration of approximately 0.01 mol/L and sodium hydroxide at a concentration of approximately 0.005 mol/L. For more information, please refer to the SDS and COA.

SRM 3180_cert

SRM 3180_SDS

Other Notes

Certified for the analytes listed below.
Iodide (I)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service