Skip to Content
Merck
All Photos(1)

Documents

NIST1954

Organic contaminants in fortified human milk

NIST® SRM® 1954

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 5 mL

manufacturer/tradename

NIST®

application(s)

food and beverages

format

matrix material

storage temp.

−20°C

General description

This Standard Reference Material (SRM) is intended for use in evaluating analytical methods for the determination of selected polychlorinated biphenyl (PCB) congeners, chlorinated pesticides, and polybrominated diphenyl ether (PBDE) congeners in human milk and similar matrices. A unit of SRM 1954 consists of five bottles of approximately 5 mL fortified human milk. For more information, please refer to the COA and SDS.

SRM 1954_cert

SRM 1954_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service