Skip to Content
Merck
All Photos(1)

Key Documents

NIST1831

Soda lime sheet glass

NIST® SRM® 1831

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Product NIST1831 is currently not available for purchase. Contact Technical Service

grade

certified reference material

Quality Level

packaging

pkg of 3 platelets

manufacturer/tradename

NIST®

application(s)

glass & ceramic

format

matrix material

General description

Each unit of SRM 1831 consists of three platelets, each measuring 37 mm square. For more information, please refer to the SDS and COA.
SRM 1831_cert

SRM 1831_SDS

Application

The Standard Reference Material (SRM) soda lime sheet glass is intended for verifying chemical analysis methods and to calibrate the instrumental analysis methods.

Features and Benefits

  • NIST-certified mass fraction values for various constituents are provided. These values are metrologically traceable to the SI unit of mass fraction and are expressed as percentage.
  • Expiration details, along with instructions for use, are included on the NIST certificate.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Silicon dioxide (SiO2), Sodium oxide (Na2O), Calcium oxide (CaO), Magnesium oxide (MgO), Aluminum Oxide (Al2O3), Potassium oxide (K2O), Sulfur trioxide (SO3), Ferric Oxide (Fe2O3), Iron(II) oxide (FeO), Titanium dioxide (TiO2)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service