Select a Size
Select a Size
About This Item
grade
certified reference material
Quality Level
packaging
pkg of disk
manufacturer/tradename
NIST®
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
SRM 1729_cert
SRM 1729_SDS
Application
Features and Benefits
- Accompanied with certificate documenting NIST-certified mass fraction values for the constituents in SRM1729.
- The reference and information values of constituents are reported as mass fraction.
- Certified values are metrologically traceable to the SI derived units of mass fraction (expressed as percent for major constituents and mg/kg for minor constituents)
- The NIST certificate is provided with period of validity, storage, safety, use, and maintenance instructions.
Other Notes
Lead (Pb), Bismuth (Bi), Zinc (Zn)
Reference/Informational value is provided for the following analytes.
Tin (Sn), Aluminium (Al), Copper (Cu), Iron (Fe), Nickel (Ni), Antimony (Sb), Silver (Ag), Silicon (Si)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service