Skip to Content
Merck
All Photos(1)

Key Documents

NIST1157

Tool steel

NIST® SRM® 1157

Sign Into View Organizational & Contract Pricing

Select a Size

1 DISC
€777.00

€777.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 DISC
€777.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€777.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

technique(s)

X-ray spectroscopy: suitable
optical emission spectroscopy (OES): suitable

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Carbon (C), Manganese (Mn), Phosphorus (P), Sulfur (S), Silicon (Si), Copper (Cu), Nickel (Ni), Chromium (Cr), Vanadium (V), Molybdenum (Mo), Tungsten (W), Cobalt (Co)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service