Skip to Content
Merck
All Photos(1)

Key Documents

N3146

Sigma-Aldrich

L-(−)-Norepinephrine (+)-bitartrate salt monohydrate

analytical standard

Synonym(s):

L-(−)-Noradrenaline (+)-bitartrate salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · C4H6O6 · H2O
CAS Number:
Molecular Weight:
337.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

packaging

vial of 200 mg

SMILES string

O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.NC[C@H](O)c1ccc(O)c(O)c1

InChI

1S/C8H11NO3.C4H6O6.H2O/c9-4-8(12)5-1-2-6(10)7(11)3-5;5-1(3(7)8)2(6)4(9)10;/h1-3,8,10-12H,4,9H2;1-2,5-6H,(H,7,8)(H,9,10);1H2/t8-;1-,2-;/m01./s1

InChI key

LNBCGLZYLJMGKP-LUDZCAPTSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in amber screw-cap vials

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service