Skip to Content
Merck
All Photos(1)

Documents

L0700000

Liothyronine sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3,3′,5-Triiodo-L-thyronine sodium salt, Liothyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine sodium salt, T3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11I3NNaO4
CAS Number:
Molecular Weight:
672.96
Beilstein:
8179867
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

liothyronine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C([O-])=O

InChI

1S/C15H12I3NO4.Na/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22;/h1-4,6,12,20H,5,19H2,(H,21,22);/q;+1/p-1/t12-;/m0./s1

InChI key

SBXXSUDPJJJJLC-YDALLXLXSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Liothyronine sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Craig M Flory et al.
The Journal of pharmacology and experimental therapeutics, 376(1), 74-83 (2020-11-01)
Acute respiratory distress syndrome (ARDS) is a severe, life-threatening form of respiratory failure characterized by pulmonary edema, inflammation, and hypoxemia due to reduced alveolar fluid clearance (AFC). Alveolar fluid clearance is required for recovery and effective gas exchange, and higher
Chen Weng et al.
Nature metabolism, 2(12), 1443-1458 (2020-12-02)
The in vitro differentiation of insulin-producing beta-like cells can model aspects of human pancreatic development. Here, we generate 95,308 single-cell transcriptomes and reconstruct a lineage tree of the entire differentiation process from human embryonic stem cells to beta-like cells to
Christopher G Hart et al.
Experimental neurology, 325, 113163-113163 (2019-12-28)
Traumatic spinal cord injury (SCI) elicits a cascade of secondary injury mechanisms that induce profound changes in glia and neurons resulting in their activation, injury or cell death. The resultant imbalanced microenvironment of acute SCI also negatively impacts regenerative processes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service