Skip to Content
Merck
All Photos(1)

Documents

IRMM428

Water (PFASs)

IRMM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

IRMM®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

JRC

format

matrix material

storage temp.

2-8°C

Analysis Note

For more information please see:
IRMM428

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Organic Pollutants: Linear perfluorooctane sulfonate (L-PFOS), Perfluorobutane sulfonate (PFBS), Perfluoroheptanoic acid (PFHpA), Perfluorohexane sulfonate (PFHxS), Perfluorohexanoic acid (PFHxA), Perfluorononanoic acid, Perfluoropentanoic acid (PFPeA)

Matrix Group: Beverages and Water

Legal Information

IRMM is a registered trademark of European Commission

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service