Skip to Content
Merck
All Photos(1)

Documents

I3132

Supelco

Indoprofen

analytical standard

Synonym(s):

α-Methyl-p-(1-oxo-2-isoindolinyl)benzeneacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15NO3
CAS Number:
Molecular Weight:
281.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CC(C(O)=O)c1ccc(cc1)N2Cc3ccccc3C2=O

InChI

1S/C17H15NO3/c1-11(17(20)21)12-6-8-14(9-7-12)18-10-13-4-2-3-5-15(13)16(18)19/h2-9,11H,10H2,1H3,(H,20,21)

InChI key

RJMIEHBSYVWVIN-UHFFFAOYSA-N

Gene Information

Application

Indoprofen is a non-steroidal drug, which can show a range of pharmacological properties such as analgesic and anti-inflammatory activities. Its mode of action involves the inhibition of prostaglandin synthesis.
Indoprofen may be used as an analytical reference standard for the quantification of the analyte in biological samples and pharmaceutical formulations using different chromatography techniques.
Indoprofen is a non-steroidal drug, which can show a range of pharmacological properties such as analgesic and anti-inflammatory activities.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kurowski et al.
International journal of clinical pharmacology research, 8(4), 287-293 (1988-01-01)
Twenty-four patients suffering from rheumatoid arthritis and requiring articular punctures were treated with 200 mg of indoprofen thrice daily for four days. The subjects were divided into four groups each of six patients. Following the last dose, blood and synovial
B Przybilla et al.
Photo-dermatology, 4(2), 73-78 (1987-04-01)
The phototoxic activity of the non-steroidal anti-inflammatory drugs (NSAID) acetylsalicylic acid, benoxaprofen, carprofen, diclofenac, indoprofen, ketoprofen, tiaprofenic acid, and of thiophene, a heterocyclic ring structure of the tiaprofenic acid molecule, was evaluated in vitro by a newly developed photo-basophil-histamine-release test
Pharmacokinetics of the enantiomers of indoprofen in man
V T, et al.
International Journal of Clinical Pharmacology Research, 4(3), 223-230 (1984)
Determination of the enantiomers of indoprofen in blood plasma by high-performance liquid chromatography after rapid derivatization by means of ethyl chloroformate
Bjorkman S
Journal of Chromatography. B, Biomedical Applications, 339(2), 339-346 (1985)
Nicholas J Lawrence et al.
The Journal of organic chemistry, 67(2), 457-464 (2002-01-19)
The intermediate anion derived from the vicarious nucleophilic substitution (VNS) of hydrogen reacts with a series of alkyl halides to generate the corresponding alpha-alkylated conventional VNS product in a one-pot process. This one-pot VNS-alkylation reaction offers a convenient route to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service