Skip to Content
Merck
All Photos(1)

Key Documents

I0328000

Iobenguane sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

m-Iodobenzylguanidine hemisulfate salt, MIBG

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€133.00

€133.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€133.00

About This Item

Linear Formula:
C8H10IN3 · 1/2H2SO4
CAS Number:
Molecular Weight:
324.13
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€133.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

iobenguane

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.NC(=N)NCc1cccc(I)c1.NC(=N)NCc2cccc(I)c2

InChI

1S/2C8H10IN3.H2O4S/c2*9-7-3-1-2-6(4-7)5-12-8(10)11;1-5(2,3)4/h2*1-4H,5H2,(H4,10,11,12);(H2,1,2,3,4)

InChI key

XNACDNPGABUBFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iobenguane sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radioiodinated metaiodobenzylguanidine (MIBG): radiochemistry, biology, and pharmacology.
Vallabhajosula S and Nikolopoulou A
Seminars in Nuclear Medicine, 41(5), 324-333 (2011)
Yan Su et al.
International journal of molecular medicine, 34(3), 842-848 (2014-06-28)
The aim of the present study was to investigate the effects of meta-iodobenzylguanidine (MIBG) on the invasive properties of hepatocellular carcinoma (HCC) cells and examine whether these effects are due to the ability of MIBG to inhibit arginine-specific mono-ADP-ribosylation. Samples
Shih-ying Huang et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 17(2), 284-294 (2014-08-26)
Iodine-131-m-iodobenzylguanidine ([(131)I]mIBG)-targeted radionuclide therapy (TRT) is a standard treatment for recurrent or refractory neuroblastoma with response rates of 30-40 %. The aim of this study is to demonstrate patient-specific dosimetry using quantitative [(124)I]mIBG positron emission tomography/X-ray computed tomography (PET/CT) imaging
Lang Rao et al.
EMBO reports, 22(2), e50613-e50613 (2020-12-22)
One major factor that contributes to the virulence of Pseudomonas aeruginosa is its ability to reside and replicate unchallenged inside airway epithelial cells. The mechanism by which P. aeruginosa escapes destruction by intracellular host defense mechanisms, such as autophagy, is not
Shohei Shinozaki et al.
Science signaling, 7(351), ra106-ra106 (2014-11-13)
Inflammation increases the abundance of inducible nitric oxide synthase (iNOS), leading to enhanced production of nitric oxide (NO), which can modify proteins by S-nitrosylation. Enhanced NO production increases the activities of the transcription factors p53 and nuclear factor κB (NF-κB)

Questions

  1. Hi, I am looking for MIBG solution with non-radioactive iodine solution which can be used for research purposes in CT scans. Could you suggest any solution? Thanks a lot.

    1 answer
    1. The I0328000 product is an analytical standard supplied by the EP. It is in powder form. Alternatively, product I9890 (also in powder form) is a research-grade option. Neither of these products would be suitable for human use and are intended for laboratory research only. See the link below to the general research-grade product:
      https://www.sigmaaldrich.com/product/sigma/i9890

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service