Skip to Content
Merck
All Photos(1)

Key Documents

H7753

Supelco

Hexestrol

analytical standard

Synonym(s):

4,4′-(1,2-Diethylethylene)diphenol, meso-3,4-Bis(4-hydroxyphenyl)hexane, Dihydrodiethylstilbestrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O2
CAS Number:
Molecular Weight:
270.37
Beilstein:
3209460
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CC[C@H]([C@H](CC)c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C18H22O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,17-20H,3-4H2,1-2H3/t17-,18+

InChI key

PBBGSZCBWVPOOL-HDICACEKSA-N

Gene Information

human ... ESR1(2099)
rat ... Esr1(24890)

Looking for similar products? Visit Product Comparison Guide

General description

Hexestrol is a synthetic nonsteroidal estrogen,[1] which can be used in human medicine as well as a growth promoter in cattle.[2]

Application

Hexestrol may be used as a reference standard for the determination of the analyte in biological samples using immunoaffinity extraction and gas chromatography-negative-ion chemical ionization mass spectrometry.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

The carcinogen hexestrol is a nonsteroidal synthetic estrogen. Metabolic activation of hexestrol to its quinone, which reacts with DNA to form analogous depurinating adducts, may be a primary critical event leading to oncogenic mutations and cancer initiation.

Linkage

Reduced analogue of diethylstilbestrol.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Todd Stravitz et al.
Clinics in liver disease, 7(2), 435-451 (2003-07-26)
Drugs rarely cause steatohepatitis, but amiodarone, perhexiline, and DH, have unequivocally been found to independently induce the histologic picture of alcoholic liver disease or NASH. All three agents have similar pathogenetic mechanisms of hepatotoxicity, targeting mitochondrial ATP production and fatty
C Logie et al.
Molecular endocrinology (Baltimore, Md.), 12(8), 1120-1132 (1998-08-26)
Activation of the estrogen receptor (ER) by hormone involves at least two steps. First, hormone binding initially relieves repression, a property imposed on ER in cis by its ligand-binding domain (EBD). Subsequently, the derepressed ER binds specific genomic sites and
A U Ziganshin et al.
Eksperimental'naia i klinicheskaia farmakologiia, 71(6), 20-22 (2009-01-15)
The results of in vitro experiments showed that hexestrol, a synthetic analog of estrogen hormones, at high concentrations inhibited the contractile response of isolated preparations of pregnant human uterus induced by ATP. It is suggested that estrogen hormones can interact
Analysis of diethylstilbestrol, dienestrol and hexestrol in biological samples by immunoaffinity extraction and gas chromatography-negative-ion chemical ionization mass spectrometry.
Bagnati R, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 527, 267-278 (1990)
Yukio Tanaka et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 45(2), 100-105 (2004-07-27)
The effects of foods and chemicals related to food hygiene on degranulation were evaluated using a method for assaying the enzyme activity of beta-hexosaminidase as an index of chemical mediator release from RBL-2H3 cells in vitro. Using a previously developed

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service