Skip to Content
Merck
All Photos(1)

Key Documents

F6145

Supelco

Fenspiride hydrochloride

analytical standard, for drug analysis

Synonym(s):

8-(2-Phenylethyl)-1-oxa-3,8-diazaspiro[4.5]decan-2-one hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20N2O2 · HCl
CAS Number:
Molecular Weight:
296.79
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

>99% (TLC)

form

powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.O=C1NCC2(CCN(CCc3ccccc3)CC2)O1

InChI

1S/C15H20N2O2.ClH/c18-14-16-12-15(19-14)7-10-17(11-8-15)9-6-13-4-2-1-3-5-13;/h1-5H,6-12H2,(H,16,18);1H

InChI key

FIKFLLIUPUVONI-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Bronchodilator with anti-inflammatory properties. Inhibits mucus secretion and reduces the release of tachykinins at a prejunctional level by its anti-muscarinic action. It also may be an antagonist at α adrenergic and H1 histamine receptors.
Bronchodilator with anti-inflammatory properties. Inhibits mucus secretion and reduces the release of tachykinins at a prejunctional level by its anti-muscarinic action. It also may be an antagonist at α adrenergic and H1 histamine receptors.
Fenspiride hydrochloride possesses anti-allergic and anti-bronchoconstrictive properties. It is used as a drug to treat inflammatory conditions of the upper and lower respiratory tracts. Experimental studies state that fenspiride has an ability to reduce the development of myringosclerosis, possibly by restricting the formation of oxygen-derived free radicals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Cortijo et al.
European journal of pharmacology, 341(1), 79-86 (1998-03-07)
We have investigated the role of human bronchial cyclic nucleotide phosphodiesterases in the effects of fenspiride, a drug endowed with bronchodilator and anti-inflammatory properties. Functional studies on human isolated bronchi showed that fenspiride (10(-6)-3 x 10(-3) M, 30 min) induced
Inhibition of the development of myringosclerosis by local administration of fenspiride, an anti-inflammatory drug.
Mattsson C and Hellstrom S
European Archives of Oto-Rhino-Laryngology : Official Journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : Affiliated With the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery, 254(9-10), 425-429 (1997)
J C Féray et al.
Biochemical pharmacology, 54(2), 293-297 (1997-07-15)
Fenspiride inhibits the calcium signal evoked by the inflammatory peptide formyl-Met-Leu-Phe (fMLP) in peritoneal macrophages, but at concentrations (approximately 1 mM) far above the therapeutic range (approximately 1 microM). Here, in rat alveolar macrophages, high fenspiride concentrations (1 mM) were
F Quartulli et al.
European journal of pharmacology, 348(2-3), 297-304 (1998-07-04)
Using the human lung epithelial WI26VA4 cell line, we investigated the capacity of fenspiride, an anti-inflammatory drug with anti-bronchoconstrictor properties, to interfere with histamine-induced intracellular Ca2+ increase and eicosanoid formation. Histamine and a histamine H1 receptor agonist elicited a rapid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service