Skip to Content
Merck
All Photos(1)

Key Documents

F0702000

Furosemide impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Chloro-4-furfurylamino-5-sulfamoylbenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€133.00

€133.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€133.00

About This Item

Empirical Formula (Hill Notation):
C12H11ClN2O5S
CAS Number:
Molecular Weight:
330.74
Beilstein:
1297887
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€133.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

furosemide

manufacturer/tradename

EDQM

mp

197 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(C1=C(Cl)C=C(NCC2=CC=CO2)C(S(N)(=O)=O)=C1)=O

InChI

1S/C12H11ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)11(21(14,18)19)4-8(9)12(16)17/h1-5,15H,6H2,(H,16,17)(H2,14,18,19)

InChI key

UXOOVYKVEXGCSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Furosemide impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service