ERMBF423A
Maize GMO Standard ERM-BF423
0% MIR604, for the detection of GMO maize (MIR604) with PCR
Synonym(s):
GVO Standard ERM-BF423, Maize MIR604
About This Item
Recommended Products
grade
certified reference material
quality
for the detection of GMO maize (MIR604) with PCR
composition
genetically modified MIR604 maize, 0%
manufacturer/tradename
JRC
application(s)
cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)
format
matrix material
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Analysis Note
ERMBF423A
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service