Skip to Content
Merck
All Photos(1)

Key Documents

ERMBF418A

Maize GMO Standard 1507

0% 1507, for the detection of GMO Maize (1507) with PCR

Synonym(s):

GMO Standard ERM-418

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€94.40

€94.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€94.40

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€94.40


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

product line

BioChemika

quality

for the detection of GMO Maize (1507) with PCR

composition

genetically modified 1507 maize, 0%

manufacturer/tradename

JRC

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
ERMBF418A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service