Skip to Content
Merck
All Photos(1)

Key Documents

ERMBC367

Rapeseed (S, total glucosinolate)

ERM®, certified reference material, high level

Sign Into View Organizational & Contract Pricing

Select a Size

20 G
€251.00

€251.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 G
€251.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

€251.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

ERM®
certified by the BSC

manufacturer/tradename

JRC

application(s)

food and beverages

format

matrix material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Rapeseed (S, total glucosinolate) has been used as a reference material in the determination of glucosinolates in horseradish plant extract using liquid chromatography coupled to high resolution mass spectrometry (LC-MS).[1]

Analysis Note

For more information please see:
ERMBC367

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: Total glucosinolate (GSL)
Trace Elements: Sulphur

Matrix Group: Nuts, Soy, Edible Oils and Fats

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucosinolates determination in tissues of horseradish Plant
Lelario F, et al.
Bio-protocol, 5 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service