Skip to Content
Merck
All Photos(1)

Key Documents

ERMBB350

Fish oil (PCB′s)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€194.00

€194.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 G
€194.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€194.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

matrix

fish

format

matrix material

Analysis Note

For more information please see:
ERMBB350

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Organic Pollutants: PCB 101, PCB 105, PCB 110, PCB 118, PCB 138, PCB 149, PCB 153, PCB 156, PCB 163, PCB 167, PCB 177, PCB 180, PCB 183, PCB 187, PCB 194, PCB 196, PCB 28, PCB 52, PCB 74, PCB 95, PCB 99

Matrix Group: Fish and Seafood

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

10 - Combustible liquids

WGK

awg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shouwei Han et al.
Molecular cancer research : MCR, 7(1), 108-117 (2009-01-17)
We previously showed that synthetic peroxisome proliferator-activated receptor gamma (PPARgamma) ligands inhibit non-small cell lung carcinoma (NSCLC) cell growth through multiple signaling pathways. Here, we show that dietary compounds, such as fish oil (which contains certain kinds of fatty acids

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service