Skip to Content
Merck
All Photos(1)

Key Documents

ERMAD455K

Creatine kinase MM isoenzyme

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€345.00

€345.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€345.00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.24

€345.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

Agency

ERM®

manufacturer/tradename

JRC

format

neat

storage temp.

−20°C

Analysis Note

For more information please see:
ERMAD455K

Legal Information

ERM is a registered trademark of European Commission

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service