Skip to Content
Merck

Skip To

E1000000

Ergometrine maleate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ergonovine maleate salt, Ergometrine maleate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H23N3O2 · C4H4O4
CAS Number:
Molecular Weight:
441.48
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

ergotamine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.C[C@@H](CO)NC(=O)[C@H]1CN(C)[C@@H]2Cc3c[nH]c4cccc(C2=C1)c34

InChI

1S/C19H23N3O2.C4H4O4/c1-11(10-23)21-19(24)13-6-15-14-4-3-5-16-18(14)12(8-20-16)7-17(15)22(2)9-13;5-3(6)1-2-4(7)8/h3-6,8,11,13,17,20,23H,7,9-10H2,1-2H3,(H,21,24);1-2H,(H,5,6)(H,7,8)/b;2-1-/t11-,13+,17+;/m0./s1

InChI key

YREISLCRUMOYAY-IIPCNOPRSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ergometrine maleate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Oxytocic, potent antagonist at smooth muscle serotonergic receptors.
Oxytocic, stimulates contraction of uterine muscle; partial agonist at serotonergic and dopaminergic receptors in CNS; selective, potent antagonist at smooth muscle serotonergic receptors.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Ergometrine maleate


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Fiorella Meneghetti et al.
Molecules (Basel, Switzerland), 25(2) (2020-01-18)
Ergometrine and methylergometrine are two alkaloids that are used as maleate salts for the prevention and control of postpartum hemorrhage. Although the two molecules have been known for a long time, few and discordant crystallographic and NMR spectroscopic data are
Jin Hee Kim et al.
Acta cardiologica, 69(6), 628-634 (2015-02-04)
Little is known about the effects of selective intracoronary administration of ergonovine into a coronary artery on the artery on the other side. This study aimed to evaluate the effect of intracoronarily administered ergonovine on the contralateral coronary artery. From



Questions

Reviews

No rating value

Active Filters