Skip to Content
Merck
All Photos(2)

Key Documents

D4065

Supelco

Dirithromycin

analytical standard, for drug analysis

Synonym(s):

[9S(R)]-9-Deoxo-11-deoxy-9,11-[imino[2-(2-methoxy)ethylidene]oxy]erthromycin; LY-237216

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H78N2O14
CAS Number:
Molecular Weight:
835.07
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard, for drug analysis

Quality Level

Assay

≥95% (TLC)

form

solid

mol wt

apparent mol wt 835.1

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)C4N[C@@H](COCCOC)OC([C@H]4C)[C@]1(C)O

InChI

1S/C42H78N2O14/c1-15-29-42(10,49)37-24(4)32(43-30(56-37)21-52-17-16-50-13)22(2)19-40(8,48)36(58-39-33(45)28(44(11)12)18-23(3)53-39)25(5)34(26(6)38(47)55-29)57-31-20-41(9,51-14)35(46)27(7)54-31/h22-37,39,43,45-46,48-49H,15-21H2,1-14H3/t22-,23-,24+,25+,26-,27+,28+,29-,30-,31+,32+,33-,34+,35+,36-,37-,39+,40-,41+,42-/m1/s1

InChI key

WLOHNSSYAXHWNR-KZYCBHIHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G G Zhanel et al.
Drugs, 61(4), 443-498 (2001-04-28)
The first macrolide, erythromycin A, demonstrated broad-spectrum antimicrobial activity and was used primarily for respiratory and skin and soft tissue infections. Newer 14-, 15- and 16-membered ring macrolides such as clarithromycin and the azalide, azithromycin, have been developed to address
In vitro activity of azithromycin and dirithromycin against axenic Entamoeba histolytica.
S Ranque et al.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology, 23(12), 932-933 (2004-12-16)
C Leroyer et al.
Respiration; international review of thoracic diseases, 65(5), 381-385 (1998-10-23)
Since a high tissue penetration of dirithromycin (D) has been assessed in early studies, the aims of this study were to determine D concentrations in bronchial mucosa and secretions in patients suffering from an acute exacerbation of chronic bronchitis (AECB)
W B Shelley et al.
Journal of the American Academy of Dermatology, 40(1), 69-72 (1999-01-28)
In an open uncontrolled study of 3 patients with balanitis xerotica obliterans we have observed significant improvement after long-term systemic antibiotic therapy. Two of the patients noticed softening of the skin as well as disappearance of pruritus, tenderness, and inflammatory
P A Moore
Journal of the American Dental Association (1939), 130(9), 1341-1343 (1999-09-24)
When treating oral infections, clinicians have used the macrolide antibiotic erythromycin as an alternative antibiotic for patients who have documented allergic reactions to penicillins. In this article, the author reports on his assessment of the pharmacology of erythromycin and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service