Skip to Content
Merck
All Photos(1)

Key Documents

D2270

Sigma-Aldrich

Dibutyl phthalate

Synonym(s):

n-Butyl phthalate, DBP, Phthalic acid dibutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
278.34
Beilstein:
1914064
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

9.6 (vs air)

vapor pressure

1 mmHg ( 147 °C)

Assay

≥98% (NLT)

form

liquid

autoignition temp.

756 °F

expl. lim.

0.47 %, 236 °F

refractive index

n20/D 1.492 (lit.)

bp

340 °C (lit.)

mp

−35 °C (lit.)

solubility

soluble, clear, colorless

density

1.043 g/mL at 25 °C (lit.)

SMILES string

CCCCOC(=O)c1ccccc1C(=O)OCCCC

InChI

1S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3

InChI key

DOIRQSBPFJWKBE-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer

Other Notes

This product has been replaced by 524980-ALDRICH | Dibutyl phthalate 99%

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

367.7 °F - open cup

Flash Point(C)

186.5 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alin C Dirtu et al.
Environment international, 59, 344-353 (2013-07-31)
Human exposure to chemicals commonly encountered in our environment, like phthalates, is routinely assessed through urinary measurement of their metabolites. A particular attention is given to the specific population groups, such as obese, for which the dietary intake of environmental
Harpreet Bhatia et al.
Environmental toxicology and chemistry, 32(10), 2335-2344 (2013-06-14)
Di-n-butyl phthalate (DnBP) is an industrial pollutant with antiandrogenic effects reported in male mammals and fish. Little research has been done on the endocrine effects of DnBP in female fish. The present study investigated the changes in ovarian histology and
Harpreet Bhatia et al.
Aquatic toxicology (Amsterdam, Netherlands), 149, 103-115 (2014-03-01)
Phthalic acid esters (PAEs) are a class of synthetic industrial chemicals commonly found in the aquatic environment worldwide. PAEs have been recognised as anti-androgens in male mammals but little is known about their endocrine disrupting effects in fish. This study
Sarah Moody et al.
Endocrinology, 154(9), 3460-3475 (2013-06-15)
Phthalates are plasticizers with widespread industrial, domestic, and medical applications. Epidemiological data indicating increased incidence of testicular dysgenesis in boys exposed to phthalates in utero are reinforced by studies demonstrating that phthalates impair fetal rodent testis development. Because humans are
Karen Sliwa et al.
Heart (British Cardiac Society), 100(24), 1967-1974 (2014-09-18)
Lack of evidence-based data on the spectrum of cardiovascular disease (CVD) in pregnancy or in the postpartum period, as well as on maternal and fetal outcome, provides challenges for treating physicians, particularly in areas of low resources. The objectives of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service