Skip to Content
Merck
All Photos(1)

Key Documents

CCW4710

Millipore

Slanetz and Bartley Agar Plates

Differential selective medium for the detection and enumeration of enterococci according to ISO Standard, suitable for microbiology

Synonym(s):

Enterococcus Selective Agar, Slanetz and Bartley Agar Plates | 55 mm plates

Sign Into View Organizational & Contract Pricing

Select a Size

30 EACH
€69.00

€69.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
30 EACH
€69.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

€69.00


Please contact Customer Service for Availability

Request more information

Agency

ISO 11133

Quality Level

form

solid

shelf life

6 mo. (Expiry date on the label)

feature

ready-to-use

composition

agar, 12.0 g/L
dipotassium phosphate, 4.00 g/L
D(+)-glucose, 2.00 g/L
sodium azide, 0.40 g/L
tryptose, 20.0 g/L
TTC, 0.10 g/L
yeast extract, 5.00 g/L

packaging

box of 30 plates

storage condition

(Store dry and tightly closed)

parameter

9 mL filling volume

plate diam.

55 mm

color

light amber to pale pink

pH

7.2±0.1 (25 °C)

application(s)

food and beverages
microbiology

storage temp.

2-25°C

suitability

Enterococcus spp.

General description

Slanetz and Bartley Agar Plates are ready-to-use plates for the enumeration and differentiation of enterococci in water samples based on the resistance to sodium azide and the ability of enterococci to reduce the TTC to formazan and so their colonies are red.

Application

Slanetz and Bartley Agar Plates are used for the enumeration and differentiation of enterococci in water samples.

Components

1 box containing: 6 plastic bags with 5 plates of 55 mm/bag

Other Notes

The color tone (light amber/pale pink) between batches can vary without modifying the characteristics of the medium.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service