Skip to Content
Merck
All Photos(1)

Key Documents

C0414

Supelco

Clidinium bromide

analytical standard

Synonym(s):

3-Hydroxy-1-methylquinuclidinium bromide benzilate, 3-[(Hydroxydiphenylacetyl)oxy]-1-methyl-1-azoniabicyclo[2.2.2]octane bromide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€82.40

€82.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€82.40

About This Item

Empirical Formula (Hill Notation):
C22H26BrNO3
CAS Number:
Molecular Weight:
432.35
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€82.40


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Br.C[N]12CCC(CC1)C(C2)OC(=O)C(O)(c3ccccc3)c4ccccc4

InChI

1S/C22H26NO3.BrH/c1-23-14-12-17(13-15-23)20(16-23)26-21(24)22(25,18-8-4-2-5-9-18)19-10-6-3-7-11-19;/h2-11,17,20,25H,12-16H2,1H3;1H

InChI key

WVDSMJSQJXGCNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Clidinium bromide is an anticholinergic, antisecretory and antispasmodic drug, widely used in treating anxiety-related conditions and spastic colon by inhibiting the action of parasympathetic innervations thus decreasing the secretions of stomach acid and slowing the intestines.[1][2]

Application

Clidinium bromide has been used as an analytical standard for the quantification of the analyte in pharmaceutical formulations[3][1][2] using different analytical techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Otto et al.
International journal of radiation applications and instrumentation. Part B, Nuclear medicine and biology, 18(5), 557-561 (1991-01-01)
The localization of quaternized muscarinic receptor (mAChR) antagonists, [11C]methyl tropanyl benzilate ([11C]MTRB) and [11C]methyl quinuclidinyl benzilate ([11C]MQNB), in rat pituitary was compared to that of [11C]tropanyl benzilate ([11C]TRB), a neutral antagonist. The quaternized ligands localize via a mAChR-mediated mechanism as
E Malatynska et al.
Brain research bulletin, 47(3), 285-290 (1998-12-29)
Three chimeric receptors stably expressed in murine fibroblast (B82) cells were used to examine how different parts of the rat muscarinic m1 and m2 receptors contribute to the down-regulation process. The MCH7 chimeric m2 receptor contained a fragment between VIth
J Delforge et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 34(6), 981-991 (1993-06-01)
The potential for noninvasive quantification of myocardial muscarinic receptors using PET data, a mathematical model, multi-injection protocols and 11C-labeled methylquinuclidinyl benzilate (MQNB) as a radioligand was previously demonstrated in dogs. The present study examines the possibility of optimizing the experimental
L Mei et al.
The Journal of pharmacology and experimental therapeutics, 251(1), 90-97 (1989-10-01)
We studied the relationship between the M1 muscarinic receptor density and the receptor-mediated hydrolysis of inositol lipids in cloned murine fibroblast B82 cells which were transfected with the m1 muscarinic receptor gene. Of the seven clones examined, the M1 muscarinic
F J Ehlert et al.
The Journal of pharmacology and experimental therapeutics, 255(3), 1148-1157 (1990-12-01)
The distribution of subtypes of the muscarinic receptor in homogenates of the rat brain was investigated by measuring the competitive inhibition of the binding [3H]N-methylscopolamine by pirenzepine and AF-DX 116 (11[[2-[(diethylamino)methyl]-1-piperidinyl]acetyl]-5, 11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepine-6-one). In most brain regions, the competitive binding curves

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service