Skip to Content
Merck
All Photos(1)

Key Documents

BP793

Budesonide

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

16,17-Butylidenebis(oxy)-11,21-dihydroxypregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing

Select a Size

110 MG
€226.00

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
110 MG
€226.00

About This Item

Empirical Formula (Hill Notation):
C25H34O6
CAS Number:
Molecular Weight:
430.53
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

budesonide

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@H]5OC(CCC)O[C@@]45C(=O)CO

InChI

1S/C25H34O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,16-18,20-22,26,28H,4-7,11-13H2,1-3H3/t16-,17-,18-,20+,21?,22+,23-,24-,25+/m0/s1

InChI key

VOVIALXJUBGFJZ-KWVAZRHASA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Budesonide BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Budesonide Aqueous Nasal Spray
  • Budesonide Inhalation Powder, pre-metered
  • Budesonide Inhalation Powder
  • Budesonide Pressurised Inhalation
  • Budesonide Nebuliser Suspension
  • Biochem/physiol Actions

    Budesonide is a second generation glucocorticoid with low systemic absorption. It is used as an anti-inflammatory agent in the treatment of asthma, rhinitis, and inflammatory bowel disease. It inhibits the expression of chemokine mRNA and production of eotaxin and RANTES protein in primary human bronchial epithelial cells. Budesonide is currently in clinical trials for the prevention of lung cancer. It shows inhibitory effects on benzo[a]pyrene-induced carcinogenesis of the lung in mice.
    Second generation glucocorticoid with low systemic absorption; anti-inflammatory agent; inhibits the expression of chemokine mRNA and production of eotaxin and RANTES protein.

    Packaging

    Unit quantity: 110 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    Health hazardExclamation mark

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Acute Tox. 4 Oral - Aquatic Chronic 3 - Repr. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

    Target Organs

    Adrenal gland

    Storage Class Code

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service