Skip to Content
Merck
All Photos(1)

Key Documents

BP762

Mercaptopurine impurity standard

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

6-Mercaptopurine monohydrate, 6-Purinethiol, 6-Thiohypoxanthine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€195.00

€195.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€195.00

About This Item

Empirical Formula (Hill Notation):
C5H4N4S·H2O
CAS Number:
Molecular Weight:
170.19
Beilstein:
4012091
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€195.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

mercaptopurine

form

solid

quality

British Pharmacopoeia (BP) Reference Standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

mixture

storage temp.

2-8°C

SMILES string

O.S=C1NC=Nc2nc[nH]c12

InChI

1S/C5H4N4S.H2O/c10-5-3-4(7-1-6-3)8-2-9-5;/h1-2H,(H2,6,7,8,9,10);1H2

InChI key

WFFQYWAAEWLHJC-UHFFFAOYSA-N

Gene Information

human ... PPAT(5471)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mercaptopurine impurity standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service