Skip to Content
Merck
All Photos(1)

Key Documents

BP725

Alverine citrate

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Alverine citrate salt, N-Ethyl-3,3′-diphenyldipropylamine, NSC 35459

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€236.00

€236.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€236.00

About This Item

Empirical Formula (Hill Notation):
C20H27N · C6H8O7
CAS Number:
Molecular Weight:
473.56
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€236.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

alverine

form

crystalline powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CCCc1ccccc1)CCCc2ccccc2

InChI

1S/C20H27N.C6H8O7/c1-2-21(17-9-15-19-11-5-3-6-12-19)18-10-16-20-13-7-4-8-14-20;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-8,11-14H,2,9-10,15-18H2,1H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

RYHCACJBKCOBTJ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Alverine citrate BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Alverine Capsules
  • Used for studies of pharmacological activity including:
    • Synergism with fluconazole against Saccharomyces cerevisiae and Candida albicans
    • Efficacy of combination with simeticone on abdominal pain associated with irritable bowel syndrome
    • Nutrient-sensitized screening for drugs that shift energy metabolism from mitochondrial respiration to glycolysis
    • Drug effects viewed from a signal transduction network perspective
    • Enhanced cytotoxic effect of proteasome inhibitor MG132
    • Mechanism of action on phasic smooth muscles

    Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service