Skip to Content
Merck
All Photos(1)

Documents

BP619

Diclofenac sodium

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Diclofenac sodium salt, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Cl2NNaO2
CAS Number:
Molecular Weight:
318.13
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diclofenac

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical (small molecule)
pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[O-]C(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl

InChI

1S/C14H11Cl2NO2.Na/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1

InChI key

KPHWPUGNDIVLNH-UHFFFAOYSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diclofenac sodium BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Diclofenac Diethylamine Gel
  • Diclofenac Prolonged-release Tablets
  • Diclofenac Gastro-resistant Tablets
  • Diclofenac Prolonged-release Capsules
  • Biochem/physiol Actions

    Standard NSAID and cyclooxygenase (COX) inhibitor. Major metabolites are 4´-hydroxydiclofenac and 5´-hydroxydiclofenac. Has been used as substrate selective for CYP2C9.

    Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    related product

    Product No.
    Description
    Pricing

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Acute Tox. 3 Oral - Aquatic Chronic 2 - Repr. 2 - STOT RE 1

    Storage Class Code

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service