Skip to Content
Merck
All Photos(1)

Key Documents

BP396

Estradiol benzoate

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

β-Estradiol 3-benzoate, 1,3,5(10)-Estratriene-3,17β-diol 3-benzoate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 3-benzoate, 3-Benzoyloxy-17β-estrol

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€194.00

€194.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€194.00

About This Item

Empirical Formula (Hill Notation):
C25H28O3
CAS Number:
Molecular Weight:
376.49
Beilstein:
3107526
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€194.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

estradiol

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

191-198 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@]3(C)[C@@H](O)CC[C@@]3([H])[C@]1([H])CCc4cc(OC(=O)c5ccccc5)ccc24

InChI

1S/C25H28O3/c1-25-14-13-20-19-10-8-18(28-24(27)16-5-3-2-4-6-16)15-17(19)7-9-21(20)22(25)11-12-23(25)26/h2-6,8,10,15,20-23,26H,7,9,11-14H2,1H3/t20-,21-,22+,23+,25+/m1/s1

InChI key

UYIFTLBWAOGQBI-BZDYCCQFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Estradiol benzoate BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.
Used in a study of general steroid metabolism and clearing, as groundwork for anabolic steroid screening in athletes.

Packaging

Unit quantity: 50 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service