Skip to Content
Merck
All Photos(1)

Key Documents

BP1103

Nicorandil

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

2-(Pyridine-3-carbonylamino)ethyl nitrate, 2-Nicotinamidoethyl nitrate, Ikorel, N-[2-(Nitrooxy)ethyl]-3-pyridinecarboxamide, SG-75, Sigmart, Zynicor

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€226.00

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€226.00

About This Item

Empirical Formula (Hill Notation):
C8H9N3O4
CAS Number:
Molecular Weight:
211.17
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€226.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

nicorandil

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)OCCNC(=O)c1cccnc1

InChI

1S/C8H9N3O4/c12-8(7-2-1-3-9-6-7)10-4-5-15-11(13)14/h1-3,6H,4-5H2,(H,10,12)

InChI key

LBHIOVVIQHSOQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nicorandil BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Nicorandil Tablets
  • Biochem/physiol Actions

    Nicorandil is a hybrid ATP-sensitive K+ (KATP) channel opener and nicotinamide nitrate NO donor.
    Nicorandil is a hybrid ATP-sensitive K+ (KATP) channel opener and nicotinamide nitrate NO donor. Nicorandil selectively activates SUR2B- versus SUR2A-containing KATP channels. It enhances endothelial NO synthase expression and protects against ischemic ventricular arrhythmias. By activating potassium channels, and donating nitric oxide to activate the enzyme guanylate cyclase, Nicorandil causes activation of GMP leading to both arterial and venous vasodilatation. Nicorandil is selective for vascular potassium channels, but has no significant action on cardiac contractility and conduction.

    Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    FlameExclamation mark

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Acute Tox. 4 Oral - Org. Perox. D

    Storage Class Code

    5.2 - Organic peroxides and self-reacting hazardous materials

    WGK

    WGK 1

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service