Skip to Content
Merck
All Photos(1)

Key Documents

BCR546

Formaldehyde-2,4-dinitrophenylhydrazone

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€196.00

€196.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€196.00

About This Item

Linear Formula:
H2C=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
210.15
Beilstein:
750330
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€196.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

mp

153-156 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(NN=C)c(c1)[N+]([O-])=O

InChI

1S/C7H6N4O4/c1-8-9-6-3-2-5(10(12)13)4-7(6)11(14)15/h2-4,9H,1H2

InChI key

UEQLSLWCHGLSML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR546

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa M Lamb et al.
PloS one, 6(11), e27149-e27149 (2011-11-17)
MAPK signal transduction pathways are important regulators of stress responses, cellular growth, and differentiation. In Neurospora, the circadian clock controls rhythms in phosphorylation of the p38-like MAPK (OS-2); however, the mechanism for this regulation is not known. We show that
Ikuo Ueta et al.
Analytical and bioanalytical chemistry, 407(3), 899-905 (2014-09-10)
A novel miniaturized sample extraction capillary was developed to provide a simple and sensitive method for analyzing gaseous formaldehyde (FA) using conventional high-performance liquid chromatography (HPLC). The extraction capillary was prepared by packing silica gel particles in a stainless steel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service