Skip to Content
Merck
All Photos(1)

Key Documents

BCR175

Tungsten (0.18 m2/g) (nitrogen BET specific surface area)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

200 G
€294.00

€294.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 G
€294.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€294.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

form

particles

manufacturer/tradename

JRC

application(s)

general analytical

format

neat

Analysis Note

For more information please see:
BCR175

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service