Skip to Content
Merck
All Photos(1)

Key Documents

BCR079R

3-Methylchrysene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€168.00

€168.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€168.00

About This Item

Empirical Formula (Hill Notation):
C19H14
CAS Number:
Molecular Weight:
242.31
Beilstein:
2557419
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€168.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

Cc1ccc2ccc3c4ccccc4ccc3c2c1

InChI

1S/C19H14/c1-13-6-7-15-9-10-17-16-5-3-2-4-14(16)8-11-18(17)19(15)12-13/h2-12H,1H3

InChI key

JLIHUJWAXSZIHS-UHFFFAOYSA-N

General description

3-Methylchrysene is a methyl derivative of chrysene, belonging to the class of polycyclic aromatic hydrocarbons (PAHs), ubiquitously present in the environment.[1] It is one of the contaminants in smoked food products.[2]

Application

3-Methylchrysene may be used as certified reference material for the quantification of the analyte in soil samples[3] and fish[4] using gas chromatography followed by mass spectrometric analysis.[3][4]

Analysis Note

For more information please see:
BCR079R

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simplified and rapid determination of polychlorinated biphenyls, polybrominated diphenyl ethers, and polycyclic aromatic hydrocarbons in fish and shrimps integrated into a single method
Kalachova K, et al.
Analytica Chimica Acta, 707(1-2), 84-91 (2011)
Occurrence of polycyclic aromatic hydrocarbons in smoked cheese
Guillen DM and Sopelana P
Journal of Dairy Science, 87(3), 556-564 (2004)
Occurrence and leachability of polycyclic aromatic compounds in contaminated soils: Chemical and bioanalytical characterization
Larsson M, et al.
The Science of the Total Environment, 622, 1476-1484 (2018)
Identification and quantification of methylated PAHs in sediment by two-dimensional gas chromatography/mass spectrometry
Skoczynska E, et al.
Analytical Methods : Advancing Methods and Applications, 5, 213-218 (2013)
Sangwoo Lee et al.
Environmental science. Processes & impacts, 19(9), 1117-1125 (2017-08-08)
Polycyclic aromatic hydrocarbons (PAHs) and alkylated PAHs are known to be major toxic contaminants in spills of petroleum hydrocarbons (oil). Spilled oil undergoes weathering and over time, PAHs go through a series of compositional changes. PAHs can disrupt endocrine functions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service