Skip to Content
Merck
All Photos(1)

Key Documents

BCR059B

Ti Al6 V4 alloy (O)

BCR®, certified reference material, cubes (0.2 g)

Synonym(s):

Titanium-aluminum-vanadium alloy, Oxygen in titanium-aluminium-vanadium alloy, Titanium-aluminum-vanadium alloy Ti with 6% Al and 4% V

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€245.00

€245.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€245.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€245.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

form

cubes (0.2 g)

manufacturer/tradename

JRC

format

matrix material

SMILES string

[Al].[Ti].[V]

InChI

1S/Al.Ti.V

InChI key

GDAKJZDZIXHMSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified for the mass fractions of nitrogen and oxygen.

Analysis Note

For more information please see:
BCR059B

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service