Skip to Content
Merck
All Photos(1)

Documents

ATG005

Sigma-Aldrich

A549 EGFRvIII Antigen Panel Cell Lines

Synonym(s):

A549 Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.41

biological source

human lung (carcinoma)

growth mode

adherent

morphology

epithelial

shipped in

dry ice

storage temp.

−196°C

General description

This product is a panel of two (2) genetically modified cell lines involving the EGFR gene and the tumor-sepcific EGFRvIII extracellular mutation. The panel consists of a EGFR knockout cell line (EGFR-/-) and an EGFRvIIIhigh expression cell line. The EGFR-/- cell line was generated using CompoZr® Zinc Finger Nuclease technology, and the EGFRvIIIhigh expression cell line was generated using MISSION® lentivirus. The EGFRvIIIhigh cell line expresses EGFRvIII at high levels relative to the EGFR-/- cell line and wild type A549 cells as measured via fluorescence-activated cell sorting (FACS).

The STR profile of these cell lines match that of its parental cell line - ECACC catalog number 86012804.

Cell Line Description

Derived from a 58 year old Caucasian male. The cells can synthesise lecithin utilising the cytidine diphosphocholine pathway. Occasional cells may also contain inclusion bodies although they are not known to carry any human pathogen.

Features and Benefits

The A549 EGFRvIII antigen panel cell lines feature the EGFRvIII extracellular mutation expressed at varying levels (KO and high). These cell lines enable researchers to screen and evaluate the efficacy and potency of EGFRvIII-targeted immunotherapies in a biologically-relevant cellular research model.

Components

ATG005A-1VL: A549 EGFRvIII Knockout
ATG005B-1VL: A549 EGFRvIII High

Quality

Tested for Mycoplasma, bacterial and fungal content, post-freeze viability, and short terminal repeat (STR) analysis for cell line identification.

Legal Information

CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service