Skip to Content
Merck
All Photos(1)

Key Documents

A7222

Sigma-Aldrich

8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt

≥96.0% purity (HPCE), solid

Synonym(s):

1,3,6-Pyrenetrisulfonic acid, 8-amino trisodium salt, APTS, Trisodium 8-aminopyrene-1,3,6-trisulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€442.00

€442.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€442.00

About This Item

Empirical Formula (Hill Notation):
C16H8NNa3O9S3
CAS Number:
Molecular Weight:
523.40
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€442.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, ≥96.0% (HPCE), solid

Quality Level

Assay

≥96.0% (HPCE)

form

solid

technique(s)

titration: suitable

color

beige to dark beige
yellow to dark yellow

mp

≥250 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].Nc1cc(c2ccc3c(cc(c4ccc1c2c34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H11NO9S3.3Na/c17-11-5-12(27(18,19)20)8-3-4-10-14(29(24,25)26)6-13(28(21,22)23)9-2-1-7(11)15(8)16(9)10;;;/h1-6H,17H2,(H,18,19,20)(H,21,22,23)(H,24,25,26);;;/q;3*+1/p-3

InChI key

XSTNYACEWLNWPY-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt (APTS) has been used for the detection of human milk oligosaccharides using multiplexed CGE with LIF (capillary gel electrophoresis with laser-induced fluorescence) technique.[1]
APTS is a fluorescence reagent that has been demonstrated to have a high level of detection sensitivity for sugars The LOD for APTS-derivatized sugars has been shown to be 0.4 nM when using a 488 nm argon-ion laser. The multi-anionic nature of APTS makes it ideal for the studies of carbohydrate molecules and small hormone molecules by high resolution capillary electrophoresis.
Fluorescent label for glycoproteins and saccharides.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a high-throughput glycoanalysis method for the characterization of oligosaccharides in human milk utilizing multiplexed capillary gel electrophoresis with laser-induced fluorescence detection.
Kottler R, et al.
Electrophoresis, 34, 2323-2336 (2013)
N N Xu et al.
Animal : an international journal of animal bioscience, 1-7 (2018-12-12)
Starchy grain is usually supplemented to diets containing low-quality forage to provide sufficient energy for ruminant animals. Ruminal degradation of grain starch mainly depends on the hydrolysis of the endosperm, which may be variable among grain sources. This study was
Alexandre Kreiss et al.
PloS one, 6(7), e22402-e22402 (2011-08-04)
Tasmanian devils (Sarcophilus harrisii) are on the verge of extinction due to a transmissible cancer, devil facial tumour disease (DFTD). This tumour is an allograft that is transmitted between individuals without immune recognition of the tumour cells. The mechanism to
Michael S Gilmore et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(23), 7273-7278 (2015-06-04)
Multidrug-resistant Enterococcus faecalis possess numerous mobile elements that encode virulence and antibiotic resistance traits as well as new metabolic pathways, often constituting over one-quarter of the genome. It was of interest to determine how this large accretion of mobile elements
Jie Chen et al.
PloS one, 10(6), e0127022-e0127022 (2015-06-16)
The aim of this study was to evaluate the diagnostic and differential diagnostic power of serum N-glycans for light chain multiple myeloma (LCMM). A total of 167 cases of subjects, including 42 LCMM, 42 IgG myeloma, 41 IgA myeloma, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service