Skip to Content
Merck
All Photos(1)

Key Documents

94610

Sigma-Aldrich

Valeroyl chloride

purum, ≥98.0% (T)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€61.70
500 ML
€169.00

€61.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€61.70
500 ML
€169.00

About This Item

Linear Formula:
CH3(CH2)3COCl
CAS Number:
Molecular Weight:
120.58
Beilstein:
506293
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€61.70


Please contact Customer Service for Availability

Request a Bulk Order

grade

purum

Quality Level

Assay

≥98.0% (T)

form

liquid

refractive index

n20/D 1.42 (lit.)
n20/D 1.420

bp

125-127 °C (lit.)

density

0.995 g/mL at 20 °C
1.016 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

CCCCC(Cl)=O

InChI

1S/C5H9ClO/c1-2-3-4-5(6)7/h2-4H2,1H3

InChI key

XGISHOFUAFNYQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janclei P Coutinho et al.
Talanta, 134, 256-263 (2015-01-27)
In this work a multivariate statistical tool (Derringer and Suich optimization) was proposed for the separation of seventeen capsinoids (natural and synthetic) using the UHPLC-DAD chromatography. Capsinoids were analyzed at 280 nm. The variables optimized were the mobile phase (water

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service