Skip to Content
Merck
All Photos(1)

Key Documents

94454

Supelco

4-(Methyl-d3-nitrosamino)-1-(3-pyridyl)-1-butanol

analytical standard

Synonym(s):

4-(Methyl-d3-nitrosoamino)-1-(3-pyridinyl)-1-butanol, NNAL-Methyl-d3

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€653.00

€653.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€653.00

About This Item

Empirical Formula (Hill Notation):
C10D3H12N3O2
CAS Number:
Molecular Weight:
212.26
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.24

€653.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

suitability

corresponds for LC-MS

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

mass shift

M+3

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])N(CCCC(O)c1cccnc1)N=O

InChI

1S/C10H15N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8,10,14H,3,5,7H2,1H3/i1D3

InChI key

OGRXKBUCZFFSTL-FIBGUPNXSA-N

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706031, T706021

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service