Skip to Content
Merck
All Photos(1)

Key Documents

94339

Sigma-Aldrich

Uridine 5′-diphosphoglucuronic acid trisodium salt

≥99.0% (HPLC)

Synonym(s):

UDP-GlcA, UDPGA, Uridine-diphosphate-glucuronic acid trisodium salt, Uridine[5′]diphospho[1]-α-D-glucopyranosuronic acid trisodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19N2Na3O18P2
CAS Number:
Molecular Weight:
646.23
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

biological source

animal

Assay

≥99.0% (HPLC)

impurities

≤10% water
≤4% solvent (ethanol)

solubility

H2O: 10 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].O[C@@H]1[C@@H](O)[C@H](O[C@@H]([C@H]1O)C([O-])=O)OP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N3C=CC(=O)NC3=O

InChI

1S/C15H22N2O18P2.3Na/c18-5-1-2-17(15(26)16-5)12-9(22)6(19)4(32-12)3-31-36(27,28)35-37(29,30)34-14-10(23)7(20)8(21)11(33-14)13(24)25;;;/h1-2,4,6-12,14,19-23H,3H2,(H,24,25)(H,27,28)(H,29,30)(H,16,18,26);;;/q;3*+1/p-3/t4-,6-,7+,8+,9-,10-,11+,12-,14-;;;/m1.../s1

InChI key

XXXUNWUNTOMVIG-QWGSIYABSA-K

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bao-Li Mi et al.
Xenobiotica; the fate of foreign compounds in biological systems, 44(8), 716-721 (2014-03-13)
1. The aurantio-obtusin's glucuronide was detected when aurantio-obtusin was incubated with human liver microsomes (HLMs). Recombinant UGT isoforms screening experiment showed that UGT1A8 was the major isoform contributed to the glucuronidation. 2. The metabolic profiles for aurantio-obtusin in liver microsomes
Jian Yu et al.
Phytotherapy research : PTR, 28(10), 1577-1580 (2014-05-21)
The present study aims to investigate the influence of irinotecan's toxicity by the biotransformation of glucoaurantio-obtusin to aurantio-obtusin. Intraperitoneal administration (i.p.) of 100 mg/kg aurantio-obtusin significantly increased the toxicity of irinotecan, but the i.p. administration of 100 mg/kg glucoaurantio-obtusin showed
Evamaria Falck et al.
Journal of pharmaceutical and biomedical analysis, 94, 36-44 (2014-02-19)
Structural modification of the GluN2B selective NMDA receptor antagonist ifenprodil led to the 3-benzazepine WMS-1410 with similar GluN2B affinity but higher receptor selectivity. Herein the in vitro and in vivo biotransformation of WMS-1410 is reported. Incubation of WMS-1410 with rat
Alexandra Maier-Salamon et al.
International journal of oncology, 45(2), 877-886 (2014-06-04)
The metabolism of 9-aminocamptothecin (9-AC) was investigated in human and rat liver microsomes. In both species 9-AC was almost exclusively biotransformed to dihydroxy-9-AC (M1) and monohydroxy-9-AC (M2). The enzymatic efficiencies of the formation of M1 and M2 (V(max)/K(m)) were 1.7-
Bo Tan et al.
Xenobiotica; the fate of foreign compounds in biological systems, 44(9), 775-784 (2014-03-19)
Leonurine is a potent component of herbal medicine Herba leonuri. The detail information on leonurine metabolism in human has not been revealed so far. Two primary metabolites, leonurine O-glucuronide and demethylated leonurine, were observed and identified in pooled human liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service